Assay Details
Gene Symbol: Rad51 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001109204.1 | NP_001102674.1 | 2 - 3 | 308 | 89 | |
XM_006234784.2 | XP_006234846.1 | 3 - 4 | 486 | 89 | ||
GenBank mRNA | BC168875.1 | - | 2 - 3 | 272 | 89 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
