Assay Details
Gene Symbol: Sptlc3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001106517.1 | NP_001099987.1 | 11 - 12 | 1825 | 111 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Agilent Whole Rat Genome Microarray 4x44K v2 | A_64_P098631 | - | - |
Agilent Whole Rat Genome Microarray 4x44K v2 | A_64_P098636 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 58546 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 703733 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 767497 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 337212 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 528111 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 879247 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 566384 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 398537 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 124018 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 687750 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 172791 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 741931 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 868201 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 479042 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 313057 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 452401 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 906433 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 965715 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 727895 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 121150 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 79978 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 484550 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 501092 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 419008 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 60844 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 207028 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 494866 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 427124 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 193569 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 801395 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 669710 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 586126 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 870336 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 838502 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 473185 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 160594 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 832258 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 327884 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 724264 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 150925 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 844332 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 458420 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 753738 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 872485 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 756924 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 201921 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 261403 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 280330 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 476975 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 314957 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 524146 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 678781 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 706743 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 503311 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 510797 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 369557 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 962234 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 488472 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 42965 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 21637 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 696231 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 462992 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 820434 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 806267 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 456314 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 512519 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 813936 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 472045 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 171886 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 706704 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 730916 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 251324 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 65795 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 175248 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 888243 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 669486 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 667810 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 890385 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 361832 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 77640 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 869345 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 219146 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 532593 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 649895 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 892349 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 250559 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 123402 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 110653 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 46606 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 141855 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 6981 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 28181 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 148098 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 161527 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 33213 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 79621 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 170392 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 3549 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 72074 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 105753 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 139761 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 152848 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 14996 | - | - |
Agilent Whole Rat Genome Microarray 4x44K v3 | A_64_P098631 | - | - |
Agilent Whole Rat Genome Microarray 4x44K v3 | A_64_P098636 | - | - |
Agilent Whole Rat Genome Microarray 4x44K v3 | A_64_P098631 | - | - |
Agilent Whole Rat Genome Microarray 4x44K v3 | A_64_P098636 | - | - |
Agilent SurePrint G3 Rat GE 8x60K Microarray | A_64_P098631 | - | - |
Agilent SurePrint G3 Rat GE 8x60K Microarray | A_64_P098636 | - | - |
SurePrint G3 Rat Gene Expression Microarray G4131A | A_44_P546128 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 33323 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 93579 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Rat Gene 1.0 ST Array | 10840276 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10840279 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10840284 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10840281 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10840283 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10840278 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10840287 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10840275 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10840286 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10840282 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10840285 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10840280 | - | - |
Affymetrix Rat Gene 1.1 ST Array | 10840274 | - | - |
Affymetrix Rat Clariom S Assay | PSR0300013227.rn.1 | TC0300001830.rn.1 | RTA TC |
Affymetrix Rat Gene 1.0 ST Array | 10840274 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
ABI Rat Genome Survey Microarray | 21995216 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Illumina ratRef-12 v1.0 expression beadchip | ILMN_1365574 | - | - |
