Assay Details
Gene Symbol: Sptlc3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001106517.1 | NP_001099987.1 | 8 - 9 | 1434 | 65 |

More Information
Set Membership: |
![]() ![]() ![]() |
Gene Ontology Categories:
