Assay Details
Gene Symbol: Mettl15 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001024981.1 | NP_001020152.1 | 3 - 4 | 459 | 57 | |
XM_006234686.2 | XP_006234748.1 | 4 - 5 | 631 | 57 | ||
XM_006234687.2 | XP_006234749.1 | 3 - 4 | 530 | 57 | ||
XM_006234688.2 | XP_006234750.1 | 4 - 5 | 626 | 57 | ||
XM_006234689.2 | XP_006234751.1 | 4 - 5 | 605 | 57 | ||
XM_008762084.1 | XP_008760306.1 | 4 - 5 | 714 | 57 | ||
XM_008762085.1 | XP_008760307.1 | 4 - 5 | 711 | 57 | ||
XM_008762086.1 | XP_008760308.1 | 4 - 5 | 693 | 57 | ||
XM_008762087.1 | XP_008760309.1 | 4 - 5 | 710 | 57 | ||
GenBank mRNA | BC091215.1 | - | 3 - 4 | 459 | 7383613 | 57 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
