Assay Details
Gene Symbol: Mettl15 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_006234686.2 | XP_006234748.1 | 6 - 7 | 1004 | 83 | |
XM_006234687.2 | XP_006234749.1 | 5 - 6 | 903 | 83 | ||
XM_006234688.2 | XP_006234750.1 | 6 - 7 | 999 | 83 | ||
XM_006234689.2 | XP_006234751.1 | 6 - 7 | 978 | 83 | ||
XM_008762084.1 | XP_008760306.1 | 6 - 7 | 1087 | 83 | ||
XM_008762085.1 | XP_008760307.1 | 6 - 7 | 1084 | 83 | ||
XM_008762086.1 | XP_008760308.1 | 6 - 7 | 1066 | 83 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Agilent Whole Rat Genome Microarray 4x44K v2 | A_64_P094060 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 680270 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 712843 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 854095 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 758308 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 233959 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 262886 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 855060 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 114318 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 351367 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 22518 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 269331 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 66951 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 45782 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 85359 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 234139 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 567221 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 912687 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 807702 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 103090 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 403371 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 917359 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 253550 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 481827 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 236026 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 968575 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 382320 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 358675 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 154424 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 813925 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 388720 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 186088 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 665795 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 654994 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 23145 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 182605 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 915780 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 463194 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 176455 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 879332 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 605025 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 855134 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 607549 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 308885 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 740326 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 393310 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 495 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 923080 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 478016 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 627692 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 758115 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 685387 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 527780 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 598509 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 699935 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 709810 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 190270 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 315826 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 503643 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 741536 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 47825 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 400619 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 11518 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 160395 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 540652 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 760342 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 312361 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 851127 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 964594 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 173387 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 234519 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 217086 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 831034 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 855980 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 915354 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 220358 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 774955 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 903609 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 595613 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 760736 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 87811 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 133978 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 117072 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 300040 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 765783 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 955653 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 300354 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 563194 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 927119 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 182292 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 929661 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 883393 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 59166 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 906061 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 423929 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 431341 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 222588 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 15841 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 330309 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 125085 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 95846 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 168590 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 67608 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 137060 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 112942 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 140547 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 115633 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 99527 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 27381 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 168947 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 46707 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 120273 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 117062 | - | - |
Agilent Whole Rat Genome Microarray 4x44K v3 | A_64_P094060 | - | - |
Agilent Whole Rat Genome Microarray 4x44K v3 | A_64_P094060 | - | - |
Agilent SurePrint G3 Rat GE 8x60K Microarray | A_64_P094060 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 102970 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 79324 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 26505 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 15561 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 104098 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 46383 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 57158 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 56793 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 11246 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 4060 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Rat Gene 1.0 ST Array | 10848023 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10848021 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10848024 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10848022 | - | - |
Affymetrix Rat Gene 1.1 ST Array | 10848019 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10848019 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
ABI Rat Genome Survey Microarray | 22106059 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Illumina ratRef-12 v1.0 expression beadchip | ILMN_1358405 | - | - |
