Assay Details
Gene Symbol: Frzb | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001100527.1 | NP_001093997.1 | 4 - 5 | 977 | 92 | |
XM_006234429.2 | XP_006234491.1 | 3 - 4 | 1401 | 92 | ||
GenBank mRNA | BC098777.1 | - | 1 - 2 | 75 | 92 | |
BC166428.1 | - | 4 - 5 | 977 | 7106784 | 92 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
