Assay Details
Gene Symbol: Frzb | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001100527.1 | NP_001093997.1 | 5 - 6 | 1174 | 99 | |
XM_006234429.2 | XP_006234491.1 | 4 - 5 | 1598 | 99 | ||
GenBank mRNA | BC098777.1 | - | 2 - 3 | 272 | 99 | |
BC166428.1 | - | 5 - 6 | 1174 | 7106784 | 99 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
