Assay Details
Gene Symbol: Cers6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_006234308.2 | XP_006234370.1 | 2 - 3 | 818 | 94 | |
XM_345363.7 | XP_345364.5 | 2 - 3 | 812 | 94 |

More Information
Set Membership: |
![]() ![]() ![]() |
Gene Ontology Categories:
