Assay Details
Gene Symbol: Prrg4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001109203.1 | NP_001102673.1 | 1 - 2 | 298 | 82 |

More Information
Additional Information:
This assay is in the 5'UTR region of the NM_001109203.1 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
