Assay Details
Gene Symbol: Prrg4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001109203.1 | NP_001102673.1 | 4 - 5 | 625 | 138 | |
XM_006234655.2 | XP_006234717.1 | 4 - 5 | 477 | 138 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
