Assay Details
Gene Symbol: Pced1a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_006235015.2 | XP_006235077.1 | 2 - 3 | 311 | 59 | |
XM_006235016.2 | XP_006235078.1 | 3 - 4 | 1683 | 59 | ||
XM_006235017.2 | XP_006235079.1 | 2 - 3 | 218 | 59 |

More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
![]() ![]() ![]() ![]() |
