Assay Details
Gene Symbol: Dnajc17 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001191740.1 | NP_001178669.1 | 1 - 2 | 104 | 76 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
