Assay Details
Gene Symbol: Pot1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001024322.1 | NP_001019493.1 | 15 - 16 | 1881 | 122 | |
XM_006236160.2 | XP_006236222.1 | 15 - 16 | 1899 | 122 | ||
XM_006236161.2 | XP_006236223.1 | 16 - 17 | 2121 | 122 | ||
XM_008762809.1 | XP_008761031.1 | 16 - 17 | 2303 | 122 | ||
XM_008762810.1 | XP_008761032.1 | 16 - 17 | 2248 | 122 | ||
XM_008762811.1 | XP_008761033.1 | 14 - 15 | 1852 | 122 | ||
GenBank mRNA | BC085834.1 | - | 15 - 16 | 1881 | 7134840 | 122 |
BC090024.1 | - | 9 - 10 | 1223 | 122 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
