Assay Details
Gene Symbol: Luc7l2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001107853.1 | NP_001101323.1 | 4 - 5 | 703 | 157 | |
XM_006236314.2 | XP_006236376.1 | 6 - 7 | 542 | 157 | ||
XM_006236315.2 | XP_006236377.1 | 5 - 6 | 1206 | 157 | ||
XM_006236316.2 | XP_006236378.1 | 5 - 6 | 435 | 157 | ||
XM_006236319.2 | XP_006236381.1 | 4 - 5 | 1135 | 157 | ||
XM_006236321.2 | XP_006236383.1 | 4 - 5 | 773 | 157 | ||
XM_006236322.2 | XP_006236384.1 | 4 - 5 | 388 | 157 | ||
XM_006236323.2 | XP_006236385.1 | 6 - 7 | 904 | 157 | ||
XM_008762778.1 | XP_008761000.1 | 5 - 6 | 3342 | 157 | ||
XM_008762779.1 | XP_008761001.1 | 5 - 6 | 3341 | 157 | ||
XM_008762780.1 | XP_008761002.1 | 5 - 6 | 642 | 157 | ||
XM_008762781.1 | XP_008761003.1 | 4 - 5 | 1155 | 157 | ||
GenBank mRNA | BC166855.1 | - | 4 - 5 | 724 | 157 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
