Assay Details
Gene Symbol: Luc7l2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001107853.1 | NP_001101323.1 | 6 - 7 | 1022 | 114 | |
XM_006236314.2 | XP_006236376.1 | 8 - 9 | 861 | 114 | ||
XM_006236315.2 | XP_006236377.1 | 7 - 8 | 1525 | 114 | ||
XM_006236316.2 | XP_006236378.1 | 7 - 8 | 754 | 114 | ||
XM_006236319.2 | XP_006236381.1 | 6 - 7 | 1454 | 114 | ||
XM_006236321.2 | XP_006236383.1 | 6 - 7 | 1092 | 114 | ||
XM_006236322.2 | XP_006236384.1 | 6 - 7 | 707 | 114 | ||
XM_006236323.2 | XP_006236385.1 | 8 - 9 | 1223 | 114 | ||
XM_008762778.1 | XP_008761000.1 | 7 - 8 | 3661 | 114 | ||
XM_008762779.1 | XP_008761001.1 | 7 - 8 | 3660 | 114 | ||
XM_008762780.1 | XP_008761002.1 | 7 - 8 | 961 | 114 | ||
XM_008762781.1 | XP_008761003.1 | 6 - 7 | 1474 | 114 | ||
GenBank mRNA | BC166855.1 | - | 6 - 7 | 1043 | 9099059 | 114 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
