Assay Details
Gene Symbol: Cnot4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001037782.1 | NP_001032871.1 | 4 - 5 | 745 | 125 | |
XM_006236271.2 | XP_006236333.1 | 4 - 5 | 763 | 125 | ||
XM_006236272.2 | XP_006236334.1 | 4 - 5 | 763 | 125 | ||
XM_006236273.2 | XP_006236335.1 | 4 - 5 | 761 | 125 | ||
XM_006236274.2 | XP_006236336.1 | 4 - 5 | 761 | 125 | ||
XM_008762777.1 | XP_008760999.1 | 4 - 5 | 765 | 125 | ||
GenBank mRNA | BC100152.1 | - | 4 - 5 | 745 | 125 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
