Assay Details
Gene Symbol: Trmt10a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001044232.1 | NP_001037697.1 | 1 - 2 | 116 | 64 | |
GenBank mRNA | BC099190.1 | - | 1 - 2 | 116 | 64 | |
FQ232089.1 | - | 1 - 2 | 132 | 64 |

More Information
Updated Mapping Information

Additional Information:
This assay is in the 5'UTR region of the BC099190.1, NM_001044232.1 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
