Assay Details
Gene Symbol: Trmt10a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001044232.1 | NP_001037697.1 | 3 - 4 | 468 | 93 | |
XM_006233368.2 | XP_006233430.1 | 3 - 4 | 2142 | 93 | ||
XM_006233369.2 | XP_006233431.1 | 3 - 4 | 688 | 93 | ||
GenBank mRNA | BC099190.1 | - | 3 - 4 | 468 | 93 | |
FQ232089.1 | - | 3 - 4 | 484 | 93 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
