Assay Details
Gene Symbol: Trmt10a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001044232.1 | NP_001037697.1 | 5 - 6 | 626 | 74 | |
XM_006233368.2 | XP_006233430.1 | 5 - 6 | 2300 | 74 | ||
XM_006233369.2 | XP_006233431.1 | 5 - 6 | 846 | 74 | ||
GenBank mRNA | BC099190.1 | - | 5 - 6 | 626 | 74 | |
FQ232089.1 | - | 5 - 6 | 642 | 74 |

More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
