Assay Details
Gene Symbol: Trmt10a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001044232.1 | NP_001037697.1 | 7 - 8 | 881 | 73 | |
XM_006233368.2 | XP_006233430.1 | 7 - 8 | 2555 | 73 | ||
XM_006233369.2 | XP_006233431.1 | 7 - 8 | 1101 | 73 | ||
GenBank mRNA | BC099190.1 | - | 7 - 8 | 881 | 73 | |
FQ232089.1 | - | 7 - 8 | 897 | 73 |

More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
