Assay Details
Gene Symbol: Henmt1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001037655.1 | NP_001032744.2 | 645 | 80 | ||
GenBank mRNA | BC107926.1 | - | 645 | 80 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
