Assay Details
Gene Symbol: Slc35a3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001012082.1 | NP_001012082.1 | 5 - 6 | 775 | 124 | |
XM_008761461.1 | XP_008759683.1 | 5 - 6 | 1043 | 124 | ||
GenBank mRNA | BC079371.1 | - | 5 - 6 | 775 | 124 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
