Assay Details
Gene Symbol: Bmpr1b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001024259.1 | NP_001019430.1 | 6 - 7 | 945 | 69 | |
XM_006233400.2 | XP_006233462.1 | 10 - 11 | 1789 | 69 | ||
XM_006233401.2 | XP_006233463.1 | 9 - 10 | 1450 | 69 | ||
XM_008761513.1 | XP_008759735.1 | 9 - 10 | 1649 | 69 | ||
XM_008761514.1 | XP_008759736.1 | 12 - 13 | 2171 | 69 | ||
GenBank mRNA | BC092609.1 | - | 6 - 7 | 945 | 7310757 | 69 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
