Assay Details
Gene Symbol: Aknad1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001044303.2 | NP_001037768.2 | 9 - 10 | 1820 | 86 | |
NR_073134.1 | 9 - 10 | 1813 | 86 | |||
XM_008761422.1 | XP_008759644.1 | 9 - 10 | 3001 | 86 | ||
GenBank mRNA | BC079051.1 | - | 9 - 10 | 1813 | 86 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
MicroArray Probes:
