Assay Details
Gene Symbol: Prkacb | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001077645.1 | NP_001071113.1 | 4 - 5 | 381 | 154 | |
XM_006233451.2 | XP_006233513.1 | 4 - 5 | 1098 | 154 | ||
XM_006233452.2 | XP_006233514.1 | 4 - 5 | 2603 | 154 | ||
XM_006233453.2 | XP_006233515.1 | 4 - 5 | 845 | 154 | ||
GenBank mRNA | D10770.1 | - | 4 - 5 | 381 | 154 |

More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
