Assay Details
Gene Symbol: Prkacb | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001077645.1 | NP_001071113.1 | 9 - 10 | 986 | 89 | |
XM_006233451.2 | XP_006233513.1 | 9 - 10 | 1703 | 89 | ||
XM_006233452.2 | XP_006233514.1 | 9 - 10 | 3208 | 89 | ||
XM_006233453.2 | XP_006233515.1 | 9 - 10 | 1450 | 89 | ||
GenBank mRNA | D10770.1 | - | 9 - 10 | 986 | 89 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
Function(s)
Process(es)
neural tube closure
protein phosphorylation
negative regulation of meiotic cell cycle
regulation of protein processing
response to clozapine
negative regulation of smoothened signaling pathway involved in dorsal/ventral neural tube patterning
magnesium ion binding
cAMP-dependent protein kinase activity
ATP binding
ubiquitin protein ligase binding
protein kinase A regulatory subunit binding
neural tube closure
protein phosphorylation
negative regulation of meiotic cell cycle
regulation of protein processing
response to clozapine
negative regulation of smoothened signaling pathway involved in dorsal/ventral neural tube patterning
magnesium ion binding
cAMP-dependent protein kinase activity
ATP binding
ubiquitin protein ligase binding
protein kinase A regulatory subunit binding
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Agilent Whole Rat Genome Microarray 4x44K v2 | A_44_P118833 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 897517 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 641711 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 433757 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 155829 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 353154 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 664960 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 729812 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 621639 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 886325 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 281083 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 473556 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 391116 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 813849 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 146957 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 73202 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 244309 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 958135 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 553015 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 312748 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 902580 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 429475 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 22847 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 420957 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 453617 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 539682 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 369728 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 227360 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 24844 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 143735 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 527404 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 611765 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 500548 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 730778 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 106827 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 527052 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 968078 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 206947 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 852569 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 241880 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 894142 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 380167 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 747168 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 364473 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 833109 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 131192 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 68272 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 62660 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 520807 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 42261 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 247020 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 831916 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 837517 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 43612 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 42968 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 424468 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 70565 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 31943 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 370175 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 660282 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 434270 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 47527 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 649919 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 863022 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 124179 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 67264 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 154823 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 816486 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 203399 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 94226 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 60956 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 165991 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 83271 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 164225 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 23243 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 143096 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 58955 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 69931 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 111241 | - | - |
Agilent Whole Rat Genome Microarray 4x44K v3 | A_44_P118833 | - | - |
Agilent Whole Rat Genome Microarray 4x44K v3 | A_44_P118833 | - | - |
Agilent SurePrint G3 Rat GE 8x60K Microarray | A_44_P118833 | - | - |
SurePrint G3 Rat Gene Expression Microarray G4131A | A_44_P880354 | - | - |
SurePrint G3 Rat Gene Expression Microarray G4131A | A_44_P158777 | - | - |
SurePrint G3 Rat Gene Expression Microarray G4131A | A_44_P745191 | - | - |
SurePrint G3 Rat Gene Expression Microarray G4131A | A_44_P402956 | - | - |
SurePrint G3 Rat Gene Expression Microarray G4131A | A_44_P118833 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 26441 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 26761 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 83343 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 20996 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Rat Gene 1.0 ST Array | 10940456 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10827308 | - | - |
Affymetrix Rat Gene 1.1 ST Array | 10827305 | - | - |
Affymetrix Rat Gene 1.1 ST Array | 10940456 | - | - |
Rat Genome 230 2.0 Array | 1371835_PM_at | - | - |
Rat Genome 230 2.0 Array | 1384772_at | - | - |
Rat Genome 230 2.0 Array | 1384772_PM_at | - | - |
Rat Genome 230 2.0 Array | 1371835_at | - | - |
Rat Genome 230 2.0 Array | 1370746_PM_at | - | - |
Rat Genome 230 2.0 Array | 1370746_at | - | - |
Affymetrix HT Rat Focus Array | 1370746_at | - | - |
Affymetrix HT Rat Focus Array | 1370746_PM_at | - | - |
Affymetrix Rat Clariom S Assay | PSR0200033754.rn.1 | TC0200004875.rn.1 | RTA TC |
Rat Expression Set 230 | 1371835_at | - | - |
Rat Expression Set 230 | 1370746_PM_at | - | - |
Rat Expression Set 230 | 1371835_PM_at | - | - |
Rat Expression Set 230 | 1370746_at | - | - |
Rat Expression Set 230 | 1384772_PM_at | - | - |
Rat Expression Set 230 | 1384772_at | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10827305 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10940456 | - | - |
Rat Genome U34 Set | D10770_s_at | - | - |
Rat Genome U34 Set | rc_AA891881_at | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
ABI Rat Genome Survey Microarray | 21330459 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Illumina ratRef-12 v1.0 expression beadchip | ILMN_1369102 | - | - |
