Assay Details
Gene Symbol: Akap8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_053855.1 | NP_446307.1 | 9 - 10 | 1177 | 85 | |
XM_006241047.2 | XP_006241109.1 | 9 - 10 | 1399 | 85 | ||
GenBank mRNA | BC166580.1 | - | 9 - 10 | 1177 | 85 | |
U01914.1 | - | 9 - 10 | 1152 | 85 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
