Assay Details
Gene Symbol: Stk38l | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001083336.1 | NP_001076805.1 | 11 - 12 | 1106 | 82 | |
XM_008763431.1 | XP_008761653.1 | 10 - 11 | 2003 | 82 | ||
GenBank mRNA | BC160812.1 | - | 9 - 10 | 988 | 82 | |
EF444939.1 | - | 11 - 12 | 1106 | 82 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
