Assay Details
Gene Symbol: Stk38l | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001083336.1 | NP_001076805.1 | 4 - 5 | 337 | 76 | |
XM_008763431.1 | XP_008761653.1 | 3 - 4 | 1234 | 76 | ||
GenBank mRNA | BC160812.1 | - | 2 - 3 | 219 | 7460150 | 76 |
EF444939.1 | - | 4 - 5 | 337 | 76 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
