Assay Details
Gene Symbol: Cd27 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001024335.1 | NP_001019506.1 | 1 - 2 | 200 | 101 | |
XM_006237380.2 | XP_006237442.1 | 1 - 2 | 1710 | 101 | ||
XM_006237381.2 | XP_006237443.1 | 1 - 2 | 1710 | 101 | ||
XM_006237382.2 | XP_006237444.1 | 1 - 2 | 1710 | 101 | ||
XM_008763293.1 | XP_008761515.1 | 2 - 3 | 1505 | 101 | ||
GenBank mRNA | BC095844.1 | - | 1 - 2 | 200 | 101 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
