Assay Details
Gene Symbol: Ppp1r42 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001127569.1 | NP_001121041.1 | 4 - 5 | 571 | 134 | |
GenBank mRNA | BC161888.1 | - | 4 - 5 | 571 | 134 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
