Assay Details
Gene Symbol: Ppp1r42 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001127569.1 | NP_001121041.1 | 8 - 9 | 1085 | 152 | |
GenBank mRNA | BC161888.1 | - | 8 - 9 | 1085 | 152 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Agilent Whole Rat Genome Microarray 4x44K v2 | A_44_P729128 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 504469 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 404438 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 81219 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 765053 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 277318 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 908732 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 458286 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 846636 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 663360 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 250491 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 100084 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 451463 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 764909 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 56095 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 850986 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 740195 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 9137 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 390300 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 614562 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 661781 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 602972 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 557047 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 427659 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 825040 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 47394 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 109643 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 705652 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 741628 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 483532 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 131282 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 69537 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 79807 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 112374 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 150256 | - | - |
Agilent Whole Rat Genome Microarray 4x44K v3 | A_44_P729128 | - | - |
Agilent Whole Rat Genome Microarray 4x44K v3 | A_44_P729128 | - | - |
Agilent SurePrint G3 Rat GE 8x60K Microarray | A_44_P729128 | - | - |
SurePrint G3 Rat Gene Expression Microarray G4131A | A_44_P729128 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 82080 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Rat Gene 1.0 ST Array | 10867313 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10867311 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10867309 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10867310 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10867308 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10867307 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10867314 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10867312 | - | - |
Affymetrix Rat Gene 1.1 ST Array | 10867306 | - | - |
Rat Genome 230 2.0 Array | 1398653_at | - | - |
Rat Genome 230 2.0 Array | 1398653_PM_at | - | - |
Affymetrix HT Rat Focus Array | 1398653_at | - | - |
Affymetrix HT Rat Focus Array | 1398653_PM_at | - | - |
Affymetrix Rat Clariom S Assay | PSR0500000549.rn.1 | TC0500000073.rn.1 | RTA TC |
Affymetrix Rat Clariom S Assay | PSR0500000550.rn.1 | TC0500000073.rn.1 | RTA TC |
Rat Expression Set 230 | 1398653_PM_at | - | - |
Rat Expression Set 230 | 1398653_at | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10867306 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
ABI Rat Genome Survey Microarray | 20920409 | - | - |
