Assay Details
Gene Symbol: Slco5a1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001107898.1 | NP_001101368.1 | 2 - 3 | 1699 | 67 | |
XM_006237743.2 | XP_006237805.1 | 3 - 4 | 1790 | 67 | ||
XM_008763495.1 | XP_008761717.1 | 3 - 4 | 2096 | 67 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
