Assay Details
Gene Symbol: Slco5a1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001107898.1 | NP_001101368.1 | 4 - 5 | 2082 | 70 | |
XM_006237743.2 | XP_006237805.1 | 5 - 6 | 2173 | 70 | ||
XM_008763495.1 | XP_008761717.1 | 5 - 6 | 2479 | 70 |

More Information
Set Membership: |
![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
