Assay Details
Gene Symbol: Terf1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001012464.1 | NP_001012482.1 | 8 - 9 | 994 | 84 | |
XM_006237729.2 | XP_006237791.1 | 7 - 8 | 967 | 84 | ||
GenBank mRNA | BC089888.1 | - | 8 - 9 | 994 | 84 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
