Assay Details
Gene Symbol: Larp4b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001107361.1 | NP_001100831.1 | 18 - 19 | 2227 | 75 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Agilent Whole Rat Genome Microarray 4x44K v2 | A_42_P705302 | - | - |
Agilent Whole Rat Genome Microarray 4x44K v2 | A_64_P123782 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 926946 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 428770 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 146999 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 473644 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 780497 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 899215 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 615452 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 213142 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 127142 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 611023 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 882071 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 531819 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 926651 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 320488 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 511484 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 195104 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 19449 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 684842 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 60178 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 768728 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 626231 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 913195 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 674428 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 661471 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 40502 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 412589 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 461758 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 93038 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 763240 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 935619 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 634179 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 67316 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 515563 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 276125 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 441647 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 682195 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 374283 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 449110 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 593493 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 173950 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 88126 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 857418 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 722579 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 317590 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 525385 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 662324 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 777623 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 162495 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 931895 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 873360 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 50811 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 186506 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 318754 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 922656 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 858613 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 8050 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 239125 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 814579 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 399268 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 778812 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 920006 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 104940 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 66450 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 11670 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 171998 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 172408 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 85468 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 84570 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 152499 | - | - |
Agilent Whole Rat Genome Microarray 4x44K v3 | A_64_P123782 | - | - |
Agilent Whole Rat Genome Microarray 4x44K v3 | A_42_P705302 | - | - |
Agilent Whole Rat Genome Microarray 4x44K v3 | A_42_P705302 | - | - |
Agilent Whole Rat Genome Microarray 4x44K v3 | A_64_P123782 | - | - |
Agilent SurePrint G3 Rat GE 8x60K Microarray | A_64_P123782 | - | - |
Agilent SurePrint G3 Rat GE 8x60K Microarray | A_42_P705302 | - | - |
SurePrint G3 Rat Gene Expression Microarray G4131A | A_44_P135610 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 66058 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 77 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Rat Gene 1.0 ST Array | 10799209 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10799221 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10799219 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10799217 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10799207 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10799202 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10799220 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10799208 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10799213 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10799218 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10799215 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10799211 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10799212 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10799216 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10799214 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10799222 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10799210 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10799201 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10799206 | - | - |
Affymetrix Rat Gene 1.1 ST Array | 10799200 | - | - |
Rat Genome 230 2.0 Array | 1391986_PM_at | - | - |
Rat Genome 230 2.0 Array | 1374879_x_at | - | - |
Rat Genome 230 2.0 Array | 1391986_at | - | - |
Rat Genome 230 2.0 Array | 1374879_PM_x_at | - | - |
Rat Genome 230 2.0 Array | 1372783_at | - | - |
Rat Genome 230 2.0 Array | 1372783_PM_at | - | - |
Affymetrix HT Rat Focus Array | 1372783_PM_at | - | - |
Affymetrix HT Rat Focus Array | 1372783_at | - | - |
Affymetrix HT Rat Focus Array | 1374879_x_at | - | - |
Affymetrix HT Rat Focus Array | 1374879_PM_x_at | - | - |
Affymetrix Rat Clariom S Assay | PSR1700005815.rn.1 | TC1700000862.rn.1 | RTA TC |
Affymetrix Rat Clariom S Assay | PSR1700005816.rn.1 | TC1700000862.rn.1 | RTA TC |
Rat Expression Set 230 | 1374879_x_at | - | - |
Rat Expression Set 230 | 1372783_at | - | - |
Rat Expression Set 230 | 1374879_PM_x_at | - | - |
Rat Expression Set 230 | 1372783_PM_at | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10799200 | - | - |
Rat Genome U34 Set | rc_AI228249_at | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
ABI Rat Genome Survey Microarray | 21695737 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Illumina ratRef-12 v1.0 expression beadchip | ILMN_1370735 | - | - |
