Assay Details
Gene Symbol: E2f2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003750061.3 | XP_003750109.1 | 1 - 2 | 807 | 67 |

More Information
Set Membership: |
![]() ![]() ![]() |
Gene Ontology Categories:
