Assay Details
Gene Symbol: E2f2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003750061.3 | XP_003750109.1 | 2 - 3 | 909 | 110 | |
XM_008764299.1 | XP_008762521.1 | 2 - 3 | 2522 | 110 |

More Information
Set Membership: |
![]() ![]() ![]() |
Gene Ontology Categories:
