Assay Details
Gene Symbol: Dhrs3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001037199.3 | NP_001032276.3 | 1 - 2 | 499 | 79 | |
GenBank mRNA | BC107450.1 | - | 1 - 2 | 499 | 7322355 | 79 |
EF125189.1 | - | 1 - 2 | 945 | 79 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
