Assay Details
Gene Symbol: Dhrs3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001037199.3 | NP_001032276.3 | 2 - 3 | 639 | 70 | |
GenBank mRNA | BC107450.1 | - | 2 - 3 | 639 | 7322355 | 70 |
EF125189.1 | - | 2 - 3 | 1085 | 70 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
