Assay Details
Gene Symbol: Ube2j2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001007655.1 | NP_001007656.1 | 2 - 3 | 494 | 68 | |
XM_006239554.2 | XP_006239616.1 | 2 - 3 | 346 | 68 | ||
XM_006239555.2 | XP_006239617.1 | 3 - 4 | 681 | 68 | ||
XM_008764352.1 | XP_008762574.1 | 2 - 3 | 504 | 68 | ||
XM_008764353.1 | XP_008762575.1 | 2 - 3 | 1574 | 68 | ||
XM_008764354.1 | XP_008762576.1 | 3 - 4 | 685 | 68 | ||
GenBank mRNA | BC079115.1 | - | 2 - 3 | 494 | 7126201 | 68 |

More Information
Additional Information:
This assay is in the 5'UTR region of the BC079115.1, NM_001007655.1 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
