Assay Details
Gene Symbol: Ube2j2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001007655.1 | NP_001007656.1 | 6 - 7 | 895 | 146 | |
XM_006239554.2 | XP_006239616.1 | 6 - 7 | 747 | 146 | ||
XM_006239555.2 | XP_006239617.1 | 7 - 8 | 1082 | 146 | ||
XM_006239557.2 | XP_006239619.1 | 5 - 6 | 742 | 146 | ||
XM_006239558.2 | XP_006239620.1 | 5 - 6 | 584 | 146 | ||
XM_008764352.1 | XP_008762574.1 | 6 - 7 | 905 | 146 | ||
XM_008764353.1 | XP_008762575.1 | 6 - 7 | 1975 | 146 | ||
XM_008764354.1 | XP_008762576.1 | 7 - 8 | 1086 | 146 | ||
XM_008764355.1 | XP_008762577.1 | 7 - 8 | 2031 | 146 | ||
GenBank mRNA | BC079115.1 | - | 6 - 7 | 895 | 146 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
