Assay Details
Gene Symbol: Dhdds | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001011978.1 | NP_001011978.1 | 1 - 2 | 76 | 108 | |
XM_006239014.2 | XP_006239076.1 | 1 - 2 | 191 | 108 | ||
GenBank mRNA | BC085875.1 | - | 1 - 2 | 76 | 108 |

More Information
Updated Mapping Information

Additional Information:
This assay is in the 5'UTR region of the BC085875.1, NM_001011978.1, XM_006239014.2 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
