Assay Details
Gene Symbol: Dhdds | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001011978.1 | NP_001011978.1 | 5 - 6 | 561 | 84 | |
XM_006239014.2 | XP_006239076.1 | 5 - 6 | 676 | 84 | ||
GenBank mRNA | BC085875.1 | - | 5 - 6 | 561 | 84 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
