Assay Details
Gene Symbol: Tti1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001134619.1 | NP_001128091.1 | 1 - 2 | 2339 | 86 | |
XM_006235428.2 | XP_006235490.1 | 2 - 3 | 2416 | 86 | ||
XM_008762375.1 | XP_008760597.1 | 2 - 3 | 3179 | 86 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
