Assay Details
Gene Symbol: Tti1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001134619.1 | NP_001128091.1 | 3 - 4 | 2687 | 97 | |
XM_006235428.2 | XP_006235490.1 | 4 - 5 | 2764 | 97 | ||
XM_008762375.1 | XP_008760597.1 | 4 - 5 | 3527 | 97 | ||
GenBank mRNA | FQ216244.1 | - | 3 - 4 | 400 | 97 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
