Assay Details
Gene Symbol: Mybl2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001106536.1 | NP_001100006.1 | 1 - 2 | 215 | 143 | |
XM_006235525.2 | XP_006235587.1 | 1 - 2 | 249 | 143 |

More Information
Additional Information:
This assay is in the 5'UTR region of the NM_001106536.1, XM_006235525.2 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
