Assay Details
Gene Symbol: Mybl2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001106536.1 | NP_001100006.1 | 5 - 6 | 694 | 77 | |
XM_006235525.2 | XP_006235587.1 | 5 - 6 | 728 | 77 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
