Assay Details
Gene Symbol: Ppp1r16b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001191072.1 | NP_001178001.1 | 3 - 4 | 500 | 94 | |
XM_008762383.1 | XP_008760605.1 | 3 - 4 | 584 | 94 | ||
XM_008762384.1 | XP_008760606.1 | 3 - 4 | 1375 | 94 | ||
XM_008762385.1 | XP_008760607.1 | 3 - 4 | 1763 | 94 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
