Assay Details
Gene Symbol: Rab5c | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001105840.2 | NP_001099310.2 | 2 - 3 | 365 | 72 | |
GenBank mRNA | BC158856.1 | - | 2 - 3 | 347 | 72 | |
CB719595.1 | - | 2 - 3 | 365 | 72 |

More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
